• USA Home
  • Search Results

  • 111trichloroethanesolution123457155611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,1-Trichloroethane solution

2 Products
Synonym:   Methyl chloroform
CAS Number:
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Purity Grade
  Package quantity
  Concentration
Show More