• USA Home
  • Search Results

  • 111trishydroxymethylpropane134177799611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,1-Tris(hydroxymethyl)propane

3 Products
Synonym:   2-Ethyl-2-hydroxymethyl-1,3-propanediol, Trimethylolpropane
Image of 1,1,1-<WBR>Tris(hydroxymethyl)<WBR>propane
CAS Number:
Linear Formula:
 CH3CH2C(CH2OH)3
Molecular Weight:
 134.17
 Beilstein/REAXYS Number:
 1698309
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More