• USA Home
  • Search Results

  • 112trichloroethane133407900511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,2-Trichloroethane

2 Products
Image of 1,1,2-<WBR>Trichloroethane
CAS Number:
Linear Formula:
 ClCH2CHCl2
Molecular Weight:
 133.40
 Beilstein/REAXYS Number:
 1731726
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More