• USA Home
  • Search Results

  • 112trichlorotrifluoroethane187387613111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,2-Trichlorotrifluoroethane

3 Products
Synonym:   1,1,2-Trichlorotrifluoroethane, CFC-113
CAS Number:
Empirical Formula (Hill Notation):
 C2Cl3F3
Molecular Weight:
 187.38
 EC Index Number:
 200-936-1
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Vapor pressure
  Transition temperature
  Solubility
  Melting Point
  Special Grade
  Vapor density vs air
  UV-Absorption Values
  Assay
Show More