• USA Home
  • Search Results

  • 113355hexamethyltrisiloxane20848118993111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,3,3,5,5-Hexamethyltrisiloxane

1 Products
Image of 1,1,3,3,5,5-<WBR>Hexamethyltrisiloxane
CAS Number:
Linear Formula:
 [(CH3)2SiHO]2Si(CH3)2
Molecular Weight:
 208.48
 Beilstein/REAXYS Number:
 1746341
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More