• USA Home
  • Search Results

  • 1133tetraethoxypropane2203112231611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,3,3-Tetraethoxypropane

1 Products
Synonym:   Malonaldehyde bis(diethyl acetal), TEP
Image of 1,1,3,3-<WBR>Tetraethoxypropane
CAS Number:
Linear Formula:
 (C2H5O)2CHCH2CH(OC2H5)2
Molecular Weight:
 220.31
 Beilstein/REAXYS Number:
 1209619
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More