• USA Home
  • Search Results

  • 1133tetramethyldisiloxane13432327726711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,3,3-Tetramethyldisiloxane

2 Products
Image of 1,1,3,3-<WBR>Tetramethyldisiloxane
CAS Number:
Linear Formula:
 [(CH3)2SiH]2O
Molecular Weight:
 134.32
 Beilstein/REAXYS Number:
 1733364
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More