• USA Home
  • Search Results

  • 1144tetraphenyl13butadiene35847145063111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,4,4-Tetraphenyl-1,3-butadiene

1 Products
Synonym:   TPB
Image of 1,1,4,4-<WBR>Tetraphenyl-<WBR>1,3-<WBR>butadiene
CAS Number:
Linear Formula:
 (C6H5)2C=CHCH=C(C6H5)2
Molecular Weight:
 358.47
 Beilstein/REAXYS Number:
 1914229
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More