• USA Home
  • Search Results

  • 11dichloro22bis4chlorophenylethene318037255911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene

1 Products
Synonym:   4,4′-DDE
Image of 1,1-<WBR>Dichloro-<WBR>2,2-<WBR>bis(4-<WBR>chlorophenyl)<WBR>ethene
CAS Number:
Linear Formula:
 (ClC6H4)2C=CCl2
Molecular Weight:
 318.03
 Beilstein/REAXYS Number:
 1913355
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More