• USA Home
  • Search Results

  • 11dichloroethylene96947535411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1-Dichloroethylene

3 Products
Synonym:   Vinylidene chloride
Image of 1,1-Dichloroethylene
CAS Number:
Linear Formula:
 CH2=CCl2
Molecular Weight:
 96.94
 Beilstein/REAXYS Number:
 1733365
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Application(s)
Show More