• USA Home
  • Search Results

  • 11mercaptoundecanoicacid218367131021911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

11-Mercaptoundecanoic acid

2 Products
Synonym:   MUA, MUDA
Image of 11-<WBR>Mercaptoundecanoic acid
CAS Number:
Linear Formula:
 HS(CH2)10CO2H
Molecular Weight:
 218.36
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More