• USA Home
  • Search Results

  • 123dichlorophenylpiperazinehydrochloride2675811953226211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(2,3-Dichlorophenyl)piperazine hydrochloride

1 Products
Synonym:   1-(2,3-dichlorophenyl)piperazine, 2,3-DCPP HCl
Image of 1-<WBR>(2,3-<WBR>Dichlorophenyl)<WBR>piperazine hydrochloride
CAS Number:
Linear Formula:
 C10H12Cl2N2 · HCl
Molecular Weight:
 267.58
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More