• USA Home
  • Search Results

  • 123trichlorobenzene181458761611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,3-Trichlorobenzene

2 Products
Image of 1,2,3-<WBR>Trichlorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H3Cl3
Molecular Weight:
 181.45
 Beilstein/REAXYS Number:
 956882
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
Show More