• USA Home
  • Search Results

  • 1245tetrachlorobenzene215899594311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,4,5-Tetrachlorobenzene

2 Products
Image of 1,2,4,5-<WBR>Tetrachlorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H2Cl4
Molecular Weight:
 215.89
 Beilstein/REAXYS Number:
 1618315
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Special Grade
  Physical Form
Show More