• USA Home
  • Search Results

  • 1245tetracyanobenzene1781571274311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,4,5-Tetracyanobenzene

1 Products
Synonym:   1,2,4,5-Benzenetetracarbonitrile, Pyromellitic acid tetranitrile
Image of 1,2,4,5-<WBR>Tetracyanobenzene
CAS Number:
Linear Formula:
 C6H2(CN)4
Molecular Weight:
 178.15
 Beilstein/REAXYS Number:
 1875027
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More