• USA Home
  • Search Results

  • 1245tetramethylbenzene134229593211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,4,5-Tetramethylbenzene

3 Products
Synonym:   1,2,4,5-Tetramethylbenzene, Durene
Image of 1,2,4,5-<WBR>Tetramethylbenzene
CAS Number:
Linear Formula:
 C6H2(CH3)4
Molecular Weight:
 134.22
 Beilstein/REAXYS Number:
 1903393
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
 eCl@ss
 39011404
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More