• USA Home
  • Search Results

  • 124triazole690728888011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,4-Triazole

2 Products
Image of 1,2,4-Triazole
CAS Number:
Empirical Formula (Hill Notation):
 C2H3N3
Molecular Weight:
 69.07
 EC Index Number
 206-022-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More