• USA Home
  • Search Results

  • 124trifluorobenzene1320836723711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2,4-Trifluorobenzene

1 Products
Image of 1,2,4-<WBR>Trifluorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H3F3
Molecular Weight:
 132.08
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More