• USA Home
  • Search Results

  • 12benzisothiazol32hone15119263433511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Benzisothiazol-3(2H)-one

2 Products
Image of 1,2-<WBR>Benzisothiazol-<WBR>3(2<I>H</I>)-one
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NOS
Molecular Weight:
 151.19
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More