• USA Home
  • Search Results

  • 12benzisoxazole1191227195411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Benzisoxazole

1 Products
Image of 1,2-Benzisoxazole
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NO
Molecular Weight:
 119.12
 Beilstein Registry Number:
 2154
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More