• USA Home
  • Search Results

  • 12bis2r5r25diethylphospholanobenzene3624713670564111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene

2 Products
Synonym:   (2R,2′R,5R,5′R)-2,2′-5,5′-Tetraethyl-1,1′-(o-phenylene)diphospholane, (R,R)-Ethyl-DUPHOS
Image of (−)-1,2-Bis[(2<I>R</I>,5<I>R</I>)-2,5-diethylphospholano]benzene
CAS Number:
Empirical Formula (Hill Notation):
 C22H36P2
Molecular Weight:
 362.47
 Beilstein/REAXYS Number:
 4814174
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More