• USA Home
  • Search Results

  • 12dibromoethane1878610693411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dibromoethane

5 Products
Synonym:   EDB, Ethylene dibromide
Image of 1,2-Dibromoethane
CAS Number:
Linear Formula:
 BrCH2CH2Br
Molecular Weight:
 187.86
 Beilstein/REAXYS Number:
 605266
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Potency
  Assay
Show More