• USA Home
  • Search Results

  • 12dibromoethanesolution1878610693411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dibromoethane solution

1 Products
Synonym:   Ethylene dibromide
Image of 1,2-Dibromoethane solution
CAS Number:
Empirical Formula (Hill Notation):
 C2H4Br2
Molecular Weight:
 187.86
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More