• USA Home
  • Search Results

  • 12dichlorobenzene147009550111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichlorobenzene

7 Products
Synonym:   o-DCB, o-dichlorobenzene
Image of 1,2-Dichlorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H4Cl2
Molecular Weight:
 147.00
 Beilstein/REAXYS Number:
 606078
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Vapor pressure
  Transition temperature
  Solubility
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Application(s)
  Product Line
  Potency
  Special Grade
  UV-Absorption Values
  Assay
Show More