• USA Home
  • Search Results

  • 12dichlorobenzened4solution15103219969111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichlorobenzene-d4 solution

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C6D4Cl2
Molecular Weight:
 151.03
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Agency/Method
  Anal. Standard Form
  Special Grade
  Package quantity
  Concentration
Show More