• USA Home
  • Search Results

  • 12dichlorobenzenesolution147009550111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichlorobenzene solution

3 Products
Image of 1,2-Dichlorobenzene solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H4Cl2
Molecular Weight:
 147.00
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Composite Dimension
  Concentration
  Special Grade
Show More