• USA Home
  • Search Results

  • 12dichloroethane989610706211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichloroethane

12 Products
Synonym:   Ethylene chloride, Ethylene dichloride, Ethylene chloride, Ethylene dichloride
Image of 1,2-Dichloroethane
CAS Number:
Empirical Formula (Hill Notation):
 C2H4Cl2
Molecular Weight:
 98.96
 EC Index Number
 203-458-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  UV-Absorption Condition
  Titertest
  Quality clarifier
  Purity Grade
  Restriction
  Transmittance
  Package quantity
  Color
  UV-Absorption Values
  Assay
Show More

Product #
Description
284505 1,2-Dichloroethane, anhydrous, 99.8%,
319929 1,2-Dichloroethane, ACS reagent, ≥99.0%,
34872 1,2-Dichloroethane, for HPLC, ≥99.8%,
02562 1,2-Dichloroethane, analytical standard,
PHR2108 1,2-Dichloroethane, Pharmaceutical Secondary Standard; Certified Reference Material,
900637 1,2-Dichloroethane, anhydrous, ZerO2®, 99.8%,
RTC000081 1,2-Dichloroethane, Pharmaceutical Secondary Standard; Certified Reference Material, New
1.13713 1,2-Dichloroethane, 1,2-Dichloroethane for liquid chromatography LiChrosolv®. CAS 107-06-2, chemical formula ClCH₂CH₂Cl.,
DX0794 1,2-Dichloroethane, anhydrous, septum-sealed bottle DriSolv®,
DX0796 1,2-Dichloroethane, OmniSolv®,
DX0800 1,2-Dichloroethane, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
1.00955 1,2-Dichloroethane, EMPLURA®,