• USA Home
  • Search Results

  • 12dichloroethaned4102981706007011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichloroethane-d4

1 Products
Synonym:   Ethylene-d4 dichloride
Image of 1,2-Dichloroethane-d<SUB>4</SUB>
CAS Number:
Linear Formula:
 ClCD2CD2Cl
Molecular Weight:
 102.98
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More