• USA Home
  • Search Results

  • 12dichloropropane112997887511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dichloropropane

3 Products
Synonym:   1,2-Dichloropropane, Propylene dichloride
Image of 1,2-Dichloropropane
CAS Number:
Linear Formula:
 CH3CHClCH2Cl
Molecular Weight:
 112.99
 Beilstein/REAXYS Number:
 1718880
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Application(s)
  Special Grade
Show More