• USA Home
  • Search Results

  • 12dimethoxyethane901211071411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dimethoxyethane

6 Products
Synonym:   DME, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme, mono-Glyme
Image of 1,2-Dimethoxyethane
CAS Number:
Linear Formula:
 CH3OCH2CH2OCH3
Molecular Weight:
 90.12
 Beilstein/REAXYS Number:
 1209237
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  pH Value
  UV-Absorption Values
  Physical Form
  Special Grade
Show More