• USA Home
  • Search Results

  • 12dimyristoylsnglycero3phosphocholine677931819424611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dimyristoyl-sn-glycero-3-phosphocholine

1 Products
Synonym:   1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 1,2-dimyristoyl, L-β,γ-Dimyristoyl-α-lecithin
Image of 1,2-Dimyristoyl-<I>sn</I>-<WBR>glycero-<WBR>3-<WBR>phosphocholine
CAS Number:
Empirical Formula (Hill Notation):
 C36H72NO8P
Molecular Weight:
 677.93
 Beilstein/REAXYS Number:
 3921768
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More