• USA Home
  • Search Results

  • 12dioleoylsnglycero3phosphorac1glycerolsodiumsalt797036725428811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

2 Products
Synonym:   (9Z)-9-Octadecenoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dioleoyl sodium salt, L-α-Phosphatidyl-DL-glycerol, dioleoyl, DOPG-Na, PG(18:1(9Z)/18:1(9Z))
Image of 1,2-Dioleoyl-<I>sn</I>-glycero-3-phospho-<I>rac</I>-(1-glycerol) sodium salt
CAS Number:
Linear Formula:
 C42H78O10PNa
Molecular Weight:
 797.03
 EC Number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More