• USA Home
  • Search Results

  • 12dipalmitoylsnglycero3phosphocholine734046389811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

2 Products
Synonym:   (7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxohexadecyl)oxy]- 3,5,9-trioxa-4-phosphapentacosan-1-aminium 4-oxide, inner salt, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, 1,2-Dihexadecanoyl-sn-glycero-3-phosphorylcholine, 1,2-Dipalmitoyl-sn-glycero-3-phosphorylcholine, 1,2-Dipalmitoyl-L-lecithin, L-α-Phosphatidylcholine, dipalmitoyl, L-β,γ-Dipalmitoyl-α-lecithin, DPPC, PC(16:0/16:0), PC
Image of 1,2-Dipalmitoyl-<I>sn</I>-<WBR>glycero-<WBR>3-<WBR>phosphocholine
CAS Number:
Empirical Formula (Hill Notation):
 C40H80NO8P
Molecular Weight:
 734.04
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Functional group
  Physical Form
  Ice Shipping
  Biological Source
Show More