• USA Home
  • Search Results

  • 12diphenoxyethane2142610466511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Diphenoxyethane

1 Products
Synonym:   Ethylene glycol diphenyl ether
Image of 1,2-Diphenoxyethane
CAS Number:
Linear Formula:
 C6H5OCH2CH2OC6H5
Molecular Weight:
 214.26
 Beilstein/REAXYS Number:
 2052248
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
140287 1,2-Diphenoxyethane, 99%,