• USA Home
  • Search Results

  • 12diphenylhydrazine1842412266711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Diphenylhydrazine

1 Products
Synonym:   N,N′-Diphenylhydrazine, N,N′-Bianiline, Hydrazobenzene, NSC 3510
Image of 1,2-<WBR>Diphenylhydrazine
CAS Number:
Linear Formula:
 C6H5NHNHC6H5
Molecular Weight:
 184.24
 Beilstein/REAXYS Number:
 639793
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More