• USA Home
  • Search Results

  • 12epoxy3phenoxypropane1501712260111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Epoxy-3-phenoxypropane

2 Products
Synonym:   2,3-Epoxypropyl phenyl ether, Glycidyl phenyl ether, Phenyl glycidyl ether
Image of 1,2-<WBR>Epoxy-<WBR>3-<WBR>phenoxypropane
CAS Number:
Empirical Formula (Hill Notation):
 C9H10O2
Molecular Weight:
 150.17
 Beilstein/REAXYS Number:
 2744
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More