• USA Home
  • Search Results

  • 12hydroxyethyl1234tetrahydro2247tetramethylquinoline233355381744011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline

1 Products
Synonym:   1,2,3,4-Tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol
Image of 1-<WBR>(2-<WBR>Hydroxyethyl)<WBR>-<WBR>1,2,3,4-<WBR>tetrahydro-<WBR>2,2,4,7-<WBR>tetramethylquinoline
CAS Number:
Empirical Formula (Hill Notation):
 C15H23NO
Molecular Weight:
 233.35
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More