• USA Home
  • Search Results

  • 12octanediol14623111786811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Octanediol

1 Products
Image of 1,2-Octanediol
CAS Number:
Linear Formula:
 CH3(CH2)5CH(OH)CH2OH
Molecular Weight:
 146.23
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
213705 1,2-Octanediol, 98%,