• USA Home
  • Search Results

  • 12propanediol76095755611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,2-Propanediol

9 Products
Synonym:   1,2-PDO, Propylene glycol
Image of 1,2-Propanediol
CAS Number:
Linear Formula:
 CH3CH(OH)CH2OH
Molecular Weight:
 76.09
 Beilstein/REAXYS Number:
 1340498
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Anal. Standard Form
  Shelf Life
  Explosion limit
  Density
  Autoignition temp
  pH Value
  Vapor pressure
  Transition temperature
  Melting Point
  Agency/Method
  Application(s)
  Potency
  Refractive Index
  Physical Form
  Special Grade
Show More