• USA Home
  • Search Results

  • 12pyridylpiperazine163223480366211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(2-Pyridyl)piperazine

2 Products
Synonym:   2-Piperazinopyridine
Image of 1-<WBR>(2-<WBR>Pyridyl)<WBR>piperazine
CAS Number:
Empirical Formula (Hill Notation):
 C9H13N3
Molecular Weight:
 163.22
 Beilstein/REAXYS Number:
 140423
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More