• USA Home
  • Search Results

  • 12trifluoromethylphenylimidazole212172537196411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-[2-(Trifluoromethyl)phenyl]imidazole

1 Products
Synonym:   TRIM
Image of 1-<WBR>[2-<WBR>(Trifluoromethyl)<WBR>phenyl]<WBR>imidazole
CAS Number:
Empirical Formula (Hill Notation):
 C10H7F3N2
Molecular Weight:
 212.17
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More