• USA Home
  • Search Results

  • 135cyclohexanetricarboxylicacid216191652668411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3,5-Cyclohexanetricarboxylic acid

2 Products
Synonym:   cis,cis-1,3,5-Cyclohexanetricarboxylic acid
Image of 1,3,5-<WBR>Cyclohexanetricarboxylic acid
CAS Number:
Linear Formula:
 C6H9(CO2H)3
Molecular Weight:
 216.19
 Beilstein/REAXYS Number:
 3120108
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
Show More