• USA Home
  • Search Results

  • 135phenyltriboronicacidtrispinacolester4560036556405211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3,5-Phenyltriboronic acid, tris(pinacol) ester

1 Products
Synonym:   1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Image of 1,3,5-<WBR>Phenyltriboronic acid, tris(pinacol) ester
CAS Number:
Empirical Formula (Hill Notation):
 C24H39B3O6
Molecular Weight:
 456.00
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More