• USA Home
  • Search Results

  • 135trifluorobenzene1320837238311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3,5-Trifluorobenzene

1 Products
Image of 1,3,5-<WBR>Trifluorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H3F3
Molecular Weight:
 132.08
 Beilstein/REAXYS Number:
 1906457
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More