• USA Home
  • Search Results

  • 135trimethoxybenzene1681962123811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3,5-Trimethoxybenzene

2 Products
Synonym:   Phloroglucinol trimethyl ether
Image of 1,3,5-<WBR>Trimethoxybenzene
CAS Number:
Linear Formula:
 C6H3(OCH3)3
Molecular Weight:
 168.19
 Beilstein/REAXYS Number:
 1307993
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Assay
Show More