• USA Home
  • Search Results

  • 135tritertbutylbenzene24643146002211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3,5-Tri-tert-butylbenzene

2 Products
Image of 1,3,5-Tri-<I>tert</I>-butylbenzene
CAS Number:
Linear Formula:
 C6H3[C(CH3)3]3
Molecular Weight:
 246.43
 Beilstein/REAXYS Number:
 1909579
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More