• USA Home
  • Search Results

  • 13benzodithiolyliumtetrafluoroborate240055784227011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Benzodithiolylium tetrafluoroborate

1 Products
Synonym:   BDTF
Image of 1,3-<WBR>Benzodithiolylium tetrafluoroborate
CAS Number:
Empirical Formula (Hill Notation):
 C7H5BF4S2
Molecular Weight:
 240.05
 Beilstein/REAXYS Number:
 4731453
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More