• USA Home
  • Search Results

  • 13butadienesolution540910699011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Butadiene solution

2 Products
Image of 1,3-Butadiene solution
CAS Number:
Empirical Formula (Hill Notation):
 C4H6
Molecular Weight:
 54.09
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Density
Show More