• USA Home
  • Search Results

  • 13chlorophenylpiperazinehydrochloride19668freebasebasis6536976811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(3-Chlorophenyl)piperazine hydrochloride

1 Products
Synonym:   m-CPP
Image of 1-<WBR>(3-<WBR>Chlorophenyl)<WBR>piperazine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H13ClN2 · xHCl
Molecular Weight:
 196.68 (free base basis)
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More