• USA Home
  • Search Results

  • 13cyclohexadiene801359257411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Cyclohexadiene

1 Products
Synonym:   1,2-Dihydrobenzene
Image of 1,3-Cyclohexadiene
CAS Number:
Empirical Formula (Hill Notation):
 C6H8
Molecular Weight:
 80.13
 Beilstein/REAXYS Number:
 506024
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More